Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo28B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144373.4 | NP_652630.2 | 3 - 4 | 1584 | 85 | |
NM_164758.2 | NP_723294.1 | 3 - 4 | 1439 | 85 | ||
NM_164759.3 | NP_723295.2 | 3 - 4 | 1439 | 85 | ||
NM_164760.4 | NP_723296.2 | 3 - 4 | 1584 | 85 | ||
GenBank mRNA | BT133443.1 | - | 3 - 4 | 1585 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |