Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259424.2 | NP_001246353.1 | 1 - 2 | 137 | 67 | |
NM_166122.4 | NP_725501.2 | 1 - 2 | 137 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |