Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140329.3 | NP_648586.2 | 1 - 2 | 1371 | 52 | |
GenBank mRNA | AY069097.1 | - | 1 - 2 | 1349 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |