Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Marcal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135039.2 | NP_608883.1 | 4 - 5 | 1649 | 127 | |
GenBank mRNA | BT011033.1 | - | 4 - 5 | 1645 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |