Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp28d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135067.3 | NP_608911.1 | 1 - 2 | 237 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |