Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pHCl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274968.1 | NP_001261897.1 | 13 | 3042 | 89 | |
NM_001274969.1 | NP_001261898.1 | 15 | 3114 | 89 | ||
NM_001274970.1 | NP_001261899.1 | 12 | 2967 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |