Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zn72D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140582.3 | NP_648839.1 | 4 - 5 | 2082 | 93 | |
GenBank mRNA | AY058357.1 | - | 4 - 5 | 2048 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |