Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140697.4 | NP_648954.1 | 2 - 3 | 1301 | 76 | |
GenBank mRNA | AB242867.1 | - | 2 - 3 | 1032 | 76 | |
AY069640.1 | - | 2 - 3 | 1256 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |