Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bru1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169492.2 | NP_001162963.1 | 5 - 6 | 1226 | 57 | |
NM_001273474.1 | NP_001260403.1 | 6 - 7 | 1864 | 57 | ||
NM_001273475.1 | NP_001260404.1 | 5 - 6 | 1098 | 57 | ||
NM_001273476.1 | NP_001260405.1 | 5 - 6 | 951 | 57 | ||
NM_001273477.1 | NP_001260406.1 | 5 - 6 | 1256 | 57 | ||
NM_001273478.1 | NP_001260407.1 | 5 - 6 | 1227 | 57 | ||
NM_165002.2 | NP_723737.1 | 6 - 7 | 2011 | 57 | ||
NM_165003.3 | NP_723738.2 | 5 - 6 | 1226 | 57 | ||
NM_165004.3 | NP_723739.1 | 5 - 6 | 1226 | 57 | ||
NM_165005.3 | NP_723740.1 | 3 - 4 | 1210 | 57 | ||
GenBank mRNA | AY051758.1 | - | 5 - 6 | 1206 | 57 | |
AY071650.1 | - | 3 - 4 | 1208 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |