Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yuri | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038821.2 | NP_001033910.2 | 4 - 5 | 875 | 61 | |
NM_001038822.3 | NP_001033911.3 | 5 - 6 | 789 | 61 | ||
NM_165106.5 | NP_723903.3 | 7 - 8 | 1661 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |