Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038817.1 | NP_001033906.1 | 8 - 9 | 1428 | 124 | |
NM_001038818.1 | NP_001033907.1 | 8 - 9 | 1601 | 124 | ||
NM_001273526.1 | NP_001260455.1 | 8 - 9 | 1674 | 124 | ||
NM_001273527.1 | NP_001260456.1 | 4 - 5 | 1021 | 124 | ||
NM_078840.5 | NP_523564.1 | 7 - 8 | 1596 | 124 | ||
NM_205988.3 | NP_995710.1 | 8 - 9 | 1692 | 124 | ||
NM_205989.3 | NP_995711.1 | 8 - 9 | 1688 | 124 | ||
GenBank mRNA | AF168467.1 | - | 8 - 9 | 1663 | 124 | |
BT010088.1 | - | 7 - 8 | 1596 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |