Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298792.1 | NP_001285721.1 | 6 - 7 | 2664 | 109 | |
NM_135309.3 | NP_609153.1 | 6 - 7 | 2664 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |