Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080366.3 | NP_525105.2 | 3 - 4 | 821 | 112 | |
NM_165394.3 | NP_724357.2 | 3 - 4 | 821 | 112 | ||
NM_165395.2 | NP_724358.2 | 3 - 4 | 869 | 112 | ||
GenBank mRNA | AY075481.1 | - | 3 - 4 | 857 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |