Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EcR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169590.2 | NP_001163061.1 | 4 - 5 | 2401 | 65 | |
NM_165461.3 | NP_724456.1 | 5 - 6 | 1640 | 65 | ||
NM_165462.2 | NP_724457.1 | 6 - 7 | 1719 | 65 | ||
NM_165463.2 | NP_724458.1 | 6 - 7 | 1765 | 65 | ||
NM_165464.3 | NP_724459.1 | 3 - 4 | 1516 | 65 | ||
NM_165465.3 | NP_724460.1 | 4 - 5 | 2482 | 65 | ||
GenBank mRNA | BT012469.1 | - | 4 - 5 | 2478 | 65 | |
BT015234.1 | - | 4 - 5 | 2477 | 65 | ||
M74078.1 | - | 4 - 5 | 2406 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |