Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136379.4 | NP_610223.1 | 2 - 3 | 530 | 65 | |
NM_165482.4 | NP_724485.1 | 2 - 3 | 530 | 65 | ||
GenBank mRNA | AY069668.1 | - | 2 - 3 | 484 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |