Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: esn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273831.1 | NP_001260760.1 | 5 - 6 | 2247 | 68 | |
NM_001273832.1 | NP_001260761.1 | 6 - 7 | 1521 | 68 | ||
NM_080251.4 | NP_524990.1 | 6 - 7 | 1831 | 68 | ||
NM_165507.2 | NP_724533.1 | 6 - 7 | 1521 | 68 | ||
GenBank mRNA | BT030763.1 | - | 6 - 7 | 1521 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |