Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299308.1 | NP_001286237.1 | 2 - 3 | 656 | 89 | |
NM_001299309.1 | NP_001286238.1 | 3 - 4 | 598 | 89 | ||
NM_080370.4 | NP_525109.1 | 3 - 4 | 2085 | 89 | ||
NM_165658.2 | NP_724763.1 | 3 - 4 | 762 | 89 | ||
NM_165659.2 | NP_724764.1 | 3 - 4 | 1661 | 89 | ||
NM_165660.2 | NP_724765.1 | 4 - 5 | 1470 | 89 | ||
NM_165663.3 | NP_724768.3 | 3 - 4 | 1125 | 89 | ||
GenBank mRNA | AY069328.1 | - | 3 - 4 | 1685 | 89 | |
BT083449.1 | - | 3 - 4 | 758 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |