Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297996.1 | NP_001284925.1 | 4 - 5 | 1309 | 64 | |
NM_001297997.1 | NP_001284926.1 | 5 - 6 | 1778 | 64 | ||
NM_078501.3 | NP_511056.2 | 4 - 5 | 1120 | 64 | ||
NM_167059.2 | NP_727065.1 | 4 - 5 | 1141 | 64 | ||
GenBank mRNA | BT003622.1 | - | 4 - 5 | 1141 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |