Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078509.3 | NP_511064.2 | 3 - 4 | 2327 | 75 | |
GenBank mRNA | BT033059.1 | - | 3 - 4 | 2292 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |