Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ork1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298167.1 | NP_001285096.1 | 4 - 5 | 1638 | 93 | |
NM_001298168.1 | NP_001285097.1 | 4 - 5 | 1208 | 93 | ||
NM_078557.5 | NP_511112.1 | 4 - 5 | 1208 | 93 | ||
NM_167259.3 | NP_727466.1 | 4 - 5 | 1329 | 93 | ||
GenBank mRNA | BT044148.1 | - | 4 - 5 | 1209 | 93 | |
U55321.1 | - | 4 - 5 | 885 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |