Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sno | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078589.4 | NP_511144.2 | 3 - 4 | 1134 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |