Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nep3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258846.1 | NP_001245775.1 | 3 - 4 | 452 | 76 | |
NM_001258847.2 | NP_001245776.1 | 3 - 4 | 590 | 76 | ||
NM_078693.3 | NP_523417.2 | 3 - 4 | 443 | 76 | ||
GenBank mRNA | BT132915.1 | - | 3 - 4 | 443 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |