Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GstE9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_166279.3 | NP_725784.1 | 1 | 622 | 61 | |
GenBank mRNA | AJ437578.1 | - | 1 | 660 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |