Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078934.3 | NP_523658.1 | 4 - 5 | 1264 | 67 | |
GenBank mRNA | AF178978.1 | - | 4 - 5 | 1249 | 67 | |
AY069670.1 | - | 4 - 5 | 1245 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |