Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136579.5 | NP_610423.3 | 4 - 5 | 1007 | 92 | |
GenBank mRNA | AY071398.1 | - | 4 - 5 | 1003 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |