Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpr47Ef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201976.2 | NP_001188905.1 | 15 - 16 | 1783 | 100 | |
NM_136816.3 | NP_610660.2 | 16 - 17 | 1816 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |