Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pds5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259342.2 | NP_001246271.1 | 13 - 14 | 4096 | 71 | |
NM_136875.4 | NP_610719.1 | 13 - 14 | 3838 | 71 | ||
GenBank mRNA | BT053744.1 | - | 13 - 14 | 3818 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |