Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmdar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014714.2 | NP_001014714.1 | 4 - 5 | 2011 | 75 | |
NM_001014715.2 | NP_001014715.1 | 3 - 4 | 1305 | 75 | ||
NM_001014716.2 | NP_001014716.1 | 3 - 4 | 962 | 75 | ||
NM_001169164.1 | NP_001162635.1 | 3 - 4 | 1019 | 75 | ||
NM_001169165.1 | NP_001162636.1 | 3 - 4 | 1029 | 75 | ||
NM_001169166.1 | NP_001162637.1 | 4 - 5 | 2012 | 75 | ||
NM_001297859.1 | NP_001284788.1 | 4 - 5 | 2012 | 75 | ||
GenBank mRNA | AY050490.1 | - | 3 - 4 | 962 | 75 | |
AY050491.1 | - | 3 - 4 | 1305 | 75 | ||
AY616144.1 | - | 3 - 4 | 1286 | 75 | ||
AY616145.1 | - | 2 - 3 | 1544 | 75 | ||
AY616146.1 | - | 3 - 4 | 1029 | 75 | ||
AY616147.1 | - | 3 - 4 | 1019 | 75 | ||
AY616148.1 | - | 4 - 5 | 2012 | 75 | ||
AY616149.1 | - | 4 - 5 | 2012 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |