Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: boi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103398.4 | NP_001096868.3 | 4 - 5 | 1568 | 68 | |
NM_001201611.2 | NP_001188540.1 | 4 - 5 | 1661 | 68 | ||
NM_001201612.2 | NP_001188541.1 | 4 - 5 | 1854 | 68 | ||
NM_166940.3 | NP_726811.1 | 4 - 5 | 1568 | 68 | ||
GenBank mRNA | BT044213.1 | - | 4 - 5 | 1528 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |