Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIP-alpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272288.2 | NP_001259217.1 | 4 - 5 | 1424 | 78 | |
NM_001272289.1 | NP_001259218.1 | 2 - 3 | 460 | 78 | ||
NM_166976.3 | NP_726871.1 | 2 - 3 | 460 | 78 | ||
GenBank mRNA | BT003528.1 | - | 2 - 3 | 457 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |