Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ArfGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079316.3 | NP_524040.2 | 4 - 5 | 1239 | 76 | |
GenBank mRNA | AY121694.1 | - | 4 - 5 | 1239 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |