Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prosbeta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079352.4 | NP_524076.2 | 1 - 2 | 184 | 73 | |
GenBank mRNA | AY051976.1 | - | 1 - 2 | 169 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |