Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ko | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275206.1 | NP_001262135.1 | 4 - 5 | 1340 | 77 | |
NM_079469.3 | NP_524193.2 | 5 - 6 | 1788 | 77 | ||
GenBank mRNA | BT150244.1 | - | 4 - 5 | 1338 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |