Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drl-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299442.1 | NP_001286371.1 | 4 - 5 | 1700 | 67 | |
NM_136991.4 | NP_610835.2 | 4 - 5 | 1700 | 67 | ||
GenBank mRNA | BT004493.1 | - | 4 - 5 | 1671 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |