Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prosap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259386.2 | NP_001246315.2 | 9 - 10 | 3448 | 85 | |
NM_001299473.1 | NP_001286402.1 | 9 - 10 | 3205 | 85 | ||
NM_001299474.1 | NP_001286403.1 | 8 - 9 | 3010 | 85 | ||
NM_137081.5 | NP_610925.3 | 10 - 11 | 3643 | 85 | ||
GenBank mRNA | BT044467.1 | - | 10 - 11 | 2726 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |