Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ih | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038856.4 | NP_001033945.1 | 12 - 13 | 5966 | 80 | |
NM_001038857.3 | NP_001033946.2 | 7 - 8 | 1361 | 80 | ||
NM_001038858.4 | NP_001033947.1 | 10 - 11 | 6678 | 80 | ||
NM_001038859.4 | NP_001033948.1 | 12 - 13 | 6035 | 80 | ||
NM_001144195.3 | NP_001137667.1 | 11 - 12 | 5939 | 80 | ||
NM_001144196.3 | NP_001137668.1 | 12 - 13 | 6020 | 80 | ||
NM_001259389.1 | NP_001246318.1 | 9 - 10 | 1463 | 80 | ||
NM_001259390.2 | NP_001246319.1 | 9 - 10 | 1502 | 80 | ||
NM_001259391.2 | NP_001246320.1 | 9 - 10 | 1504 | 80 | ||
GenBank mRNA | AY928070.1 | - | 12 - 13 | 3427 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |