Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: regucalcin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132547.3 | NP_572775.1 | 2 - 3 | 811 | 91 | |
NM_167314.2 | NP_727586.1 | 2 - 3 | 865 | 91 | ||
NM_167315.2 | NP_727587.1 | 2 - 3 | 855 | 91 | ||
NM_167316.2 | NP_727588.2 | 2 - 3 | 797 | 91 | ||
GenBank mRNA | AB036904.1 | - | 2 - 3 | 894 | 91 | |
AY118643.1 | - | 2 - 3 | 790 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |