Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bgm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298994.1 | NP_001285923.1 | 4 - 5 | 1235 | 62 | |
NM_079959.4 | NP_524698.1 | 4 - 5 | 1272 | 62 | ||
GenBank mRNA | BT044224.1 | - | 4 - 5 | 1253 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |