Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp52F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137249.4 | NP_611093.2 | 3 - 4 | 427 | 96 | |
GenBank mRNA | BT133387.1 | - | 3 - 4 | 407 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |