Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp52F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137249.4 | NP_611093.2 | 7 - 8 | 1497 | 74 | |
GenBank mRNA | BT133387.1 | - | 7 - 8 | 1477 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |