Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mamo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201660.3 | NP_001188589.1 | 5 - 6 | 3110 | 96 | |
NM_001258732.2 | NP_001245661.1 | 7 - 8 | 3971 | 96 | ||
NM_001258733.1 | NP_001245662.1 | 6 - 7 | 3095 | 96 | ||
NM_001272610.2 | NP_001259539.1 | 7 - 8 | 3985 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |