Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RhoGEF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144390.2 | NP_001137862.1 | 5 - 6 | 1892 | 80 | |
NM_001144391.1 | NP_001137863.1 | 3 - 4 | 1805 | 80 | ||
NM_001169836.1 | NP_001163307.1 | 5 - 6 | 1629 | 80 | ||
NM_001169837.1 | NP_001163308.1 | 5 - 6 | 1704 | 80 | ||
NM_001274273.1 | NP_001261202.1 | 14 - 15 | 9658 | 80 | ||
NM_001382119.1 | NP_001368935.1 | 4 - 5 | 1756 | 80 | ||
NM_138180.4 | NP_612024.4 | 14 - 15 | 9661 | 80 | ||
NM_138182.2 | NP_612026.2 | 5 - 6 | 2048 | 80 | ||
NM_138183.3 | NP_612027.2 | 5 - 6 | 1892 | 80 | ||
NM_167823.2 | NP_728515.1 | 5 - 6 | 1721 | 80 | ||
NM_167824.2 | NP_728516.1 | 5 - 6 | 1715 | 80 | ||
NM_167825.2 | NP_728517.1 | 4 - 5 | 1700 | 80 | ||
GenBank mRNA | BT044233.1 | - | 5 - 6 | 1872 | 80 | |
BT099707.1 | - | 5 - 6 | 2048 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |