Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299259.1 | NP_001286188.1 | 5 - 6 | 2416 | 105 | |
NM_143713.3 | NP_651970.1 | 4 - 5 | 2273 | 105 | ||
GenBank mRNA | BT004908.1 | - | 4 - 5 | 2233 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |