Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcyt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139365.3 | NP_647622.1 | 1 - 2 | 519 | 73 | |
GenBank mRNA | AY058458.1 | - | 1 - 2 | 519 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |