Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scramb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139504.3 | NP_647761.1 | 3 - 4 | 833 | 89 | |
GenBank mRNA | AY060271.1 | - | 3 - 4 | 907 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |