Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Argk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104086.2 | NP_001097556.2 | 2 - 3 | 625 | 86 | |
NM_168313.4 | NP_729448.1 | 1 - 2 | 767 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |