Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fry | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259770.1 | NP_001246699.1 | 19 - 20 | 6314 | 71 | |
NM_001259771.1 | NP_001246700.1 | 18 - 19 | 5811 | 71 | ||
NM_001259772.1 | NP_001246701.1 | 19 - 20 | 5955 | 71 | ||
NM_001274708.1 | NP_001261637.1 | 18 - 19 | 6170 | 71 | ||
NM_001274709.1 | NP_001261638.1 | 19 - 20 | 5955 | 71 | ||
NM_001274710.1 | NP_001261639.1 | 18 - 19 | 5811 | 71 | ||
NM_168356.2 | NP_729519.2 | 19 - 20 | 5955 | 71 | ||
NM_168357.3 | NP_729520.1 | 18 - 19 | 5811 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |