Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ru | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080051.3 | NP_524790.1 | 3 - 4 | 1030 | 98 | |
GenBank mRNA | AF318284.1 | - | 3 - 4 | 1035 | 98 | |
AY118997.1 | - | 3 - 4 | 1029 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |