Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGRP-SA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298199.1 | NP_001285128.1 | 1 - 2 | 345 | 77 | |
NM_132499.3 | NP_572727.1 | 1 - 2 | 637 | 77 | ||
GenBank mRNA | AF207541.1 | - | 1 - 2 | 314 | 77 | |
AY075293.1 | - | 1 - 2 | 752 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster Inventoried |