Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RhoGAP15B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042817.3 | NP_001036282.1 | 7 - 8 | 4492 | 67 | |
NM_132955.4 | NP_573183.2 | 7 - 8 | 4618 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |